Assay Details
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717707.3 | XP_002717753.1 | 4 - 5 | 543 | 92 | |
XM_008268955.1 | XP_008267177.1 | 5 - 6 | 622 | 92 | ||
XM_008268956.2 | XP_008267178.1 | 5 - 6 | 624 | 92 | ||
XM_008268957.2 | XP_008267179.1 | 4 - 5 | 544 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
