Assay Details
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717707.3 | XP_002717753.1 | 7 - 8 | 934 | 99 | |
XM_008268955.1 | XP_008267177.1 | 8 - 9 | 1013 | 99 | ||
XM_008268956.2 | XP_008267178.1 | 8 - 9 | 1015 | 99 | ||
XM_008268957.2 | XP_008267179.1 | 7 - 8 | 935 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
