Assay Details
Gene Symbol: GNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712904.3 | XP_002712950.1 | 4 - 5 | 1321 | 65 | |
XM_017343413.1 | XP_017198902.1 | 4 - 5 | 1257 | 65 | ||
XM_017343414.1 | XP_017198903.1 | 3 - 4 | 1234 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
