Assay Details
Gene Symbol: DNER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008259280.1 | XP_008257502.1 | 11 - 12 | 1979 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DNER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008259280.1 | XP_008257502.1 | 11 - 12 | 1979 | 52 |
Set Membership: |
![]() ![]() ![]() ![]() |