Assay Details
Gene Symbol: TXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711412.3 | XP_002711458.1 | 1 - 2 | 112 | 119 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002711412.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
