Assay Details
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008254371.1 | XP_008252593.1 | 2 - 3 | 518 | 77 | |
XM_017340749.1 | XP_017196238.1 | 2 - 3 | 518 | 77 | ||
XM_017340750.1 | XP_017196239.1 | 2 - 3 | 518 | 77 | ||
XM_017340751.1 | XP_017196240.1 | 2 - 3 | 518 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
