Assay Details
Gene Symbol: WDR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002721720.3 | XP_002721766.1 | 473 | 99 | ||
XM_008248670.2 | XP_008246892.1 | 473 | 99 | |||
XM_008248671.2 | XP_008246893.1 | 473 | 99 | |||
XM_008248672.2 | XP_008246894.1 | 473 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
