Assay Details
Gene Symbol: LOC103351479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017349121.1 | XP_017204610.1 | 1 - 2 | 170 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC103351479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017349121.1 | XP_017204610.1 | 1 - 2 | 170 | 89 |
Set Membership: |
![]() ![]() ![]() ![]() |