Assay Details
Gene Symbol: HPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008270316.2 | XP_008268538.1 | 4 - 5 | 662 | 60 | |
XM_008270317.1 | XP_008268539.1 | 4 - 5 | 500 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
