Assay Details
Gene Symbol: TRAF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709054.3 | XP_002709100.1 | 6 - 7 | 1036 | 58 | |
XM_017346011.1 | XP_017201500.1 | 6 - 7 | 986 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
