Assay Details
Gene Symbol: TENT4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002721816.3 | XP_002721862.1 | 1606 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
