Assay Details
Gene Symbol: MOB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723758.3 | XP_002723804.1 | 359 | 80 | ||
XM_017339850.1 | XP_017195339.1 | 189 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017339850.1 transcripts.
Set Membership: |
![]() ![]() |
