Assay Details
Gene Symbol: TAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017349836.1 | XP_017205325.1 | 7 - 8 | 2390 | 87 | |
XM_017349837.1 | XP_017205326.1 | 7 - 8 | 2342 | 87 | ||
XM_017349838.1 | XP_017205327.1 | 6 - 7 | 2314 | 87 | ||
XM_017349839.1 | XP_017205328.1 | 6 - 7 | 2266 | 87 | ||
XM_017349840.1 | XP_017205329.1 | 4 - 5 | 2108 | 87 | ||
XM_017349841.1 | XP_017205330.1 | 7 - 8 | 2394 | 87 | ||
XM_017349842.1 | XP_017205331.1 | 4 - 5 | 1855 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
