Assay Details
Gene Symbol: THPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266592.2 | XP_008264814.1 | 1 - 2 | 111 | 62 | |
XM_008266593.2 | XP_008264815.1 | 1 - 2 | 111 | 62 | ||
XM_008266594.2 | XP_008264816.1 | 1 - 2 | 111 | 62 | ||
XM_017346953.1 | XP_017202442.1 | 2 - 3 | 308 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008266592.2, XM_008266593.2, XM_008266594.2, XM_017346953.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
