Assay Details
Gene Symbol: CENPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262306.2 | XP_008260528.1 | 2 - 3 | 254 | 90 | |
XM_008262307.2 | XP_008260529.1 | 2 - 3 | 462 | 90 | ||
XM_008262308.2 | XP_008260530.1 | 2 - 3 | 254 | 90 | ||
XM_008262309.2 | XP_008260531.1 | 2 - 3 | 254 | 90 | ||
XM_017344765.1 | XP_017200254.1 | 2 - 3 | 254 | 90 | ||
XM_017344766.1 | XP_017200255.1 | 2 - 3 | 254 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008262307.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
