Assay Details
Gene Symbol: BCL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722735.3 | XP_002722781.1 | 292 | 82 |

More Information
Set Membership: |
![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BCL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722735.3 | XP_002722781.1 | 292 | 82 |
Set Membership: |
![]() ![]() |