Assay Details
Gene Symbol: PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249596.2 | XP_008247818.1 | 207 | 105 | ||
XM_008249597.2 | XP_008247819.1 | 197 | 105 | |||
XM_008249598.2 | XP_008247820.1 | 262 | 105 |

More Information
Set Membership: |
![]() ![]() |
