Assay Details
Gene Symbol: PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249596.2 | XP_008247818.1 | 336 | 80 | ||
XM_008249597.2 | XP_008247819.1 | 326 | 80 | |||
XM_008249598.2 | XP_008247820.1 | 391 | 80 |

More Information
Set Membership: |
![]() ![]() |
