Assay Details
Gene Symbol: MYOZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008255266.2 | XP_008253488.1 | 3 - 4 | 839 | 71 | |
XM_008255267.2 | XP_008253489.1 | 4 - 5 | 890 | 71 | ||
XM_008255268.2 | XP_008253490.1 | 4 - 5 | 913 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
