Assay Details
Gene Symbol: CALCOCO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008271338.2 | XP_008269560.1 | 2 - 3 | 297 | 61 | |
XM_008271339.1 | XP_008269561.1 | 2 - 3 | 298 | 61 | ||
XM_017349164.1 | XP_017204653.1 | 2 - 3 | 297 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
