Assay Details
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716781.3 | XP_002716827.1 | 2 - 3 | 240 | 65 | |
XM_008267166.2 | XP_008265388.1 | 2 - 3 | 350 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
