Assay Details
Gene Symbol: UMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017344468.1 | XP_017199957.1 | 4 - 5 | 546 | 103 | |
XM_017344469.1 | XP_017199958.1 | 3 - 4 | 210 | 103 | ||
XM_017344470.1 | XP_017199959.1 | 3 - 4 | 344 | 103 | ||
XM_017344471.1 | XP_017199960.1 | 3 - 4 | 358 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
