Assay Details
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017350354.1 | XP_017205843.1 | 3890 | 141 | ||
XM_017350355.1 | XP_017205844.1 | 3891 | 141 | |||
XM_017350356.1 | XP_017205845.1 | 3744 | 141 |

More Information
Set Membership: |
![]() ![]() |
