Assay Details
Gene Symbol: DMAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249693.2 | XP_008247915.2 | 457 | 76 |

More Information
Set Membership: |
![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DMAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249693.2 | XP_008247915.2 | 457 | 76 |
Set Membership: |
![]() ![]() |