Assay Details
Gene Symbol: LOC100349431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717918.3 | XP_002717964.1 | 2 - 3 | 829 | 55 | |
XM_008269433.2 | XP_008267655.1 | 2 - 3 | 829 | 55 | ||
XM_008269434.2 | XP_008267656.1 | 2 - 3 | 829 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
