Assay Details
Gene Symbol: ADNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008250426.2 | XP_008248648.1 | 315 | 59 | ||
XM_008250427.2 | XP_008248649.1 | 1871 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
