Assay Details
Gene Symbol: WBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008254214.2 | XP_008252436.2 | 4 - 5 | 610 | 78 | |
XM_008254215.2 | XP_008252437.2 | 4 - 5 | 522 | 78 | ||
XM_008254216.2 | XP_008252438.2 | 3 - 4 | 419 | 78 | ||
XM_017340590.1 | XP_017196079.1 | 4 - 5 | 699 | 78 | ||
XM_017340591.1 | XP_017196080.1 | 4 - 5 | 751 | 78 | ||
XM_017340592.1 | XP_017196081.1 | 3 - 4 | 510 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
