Assay Details
Gene Symbol: ATXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720866.3 | XP_002720912.1 | 711 | 102 | ||
XM_008274135.2 | XP_008272357.1 | 844 | 102 | |||
XM_008274137.2 | XP_008272359.1 | 906 | 102 | |||
XM_008274138.2 | XP_008272360.1 | 773 | 102 | |||
XM_017350442.1 | XP_017205931.1 | 673 | 102 | |||
XM_017350444.1 | XP_017205933.1 | 646 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002720866.3, XM_008274135.2, XM_008274137.2, XM_008274138.2, XM_017350442.1, XM_017350444.1 transcripts.
Set Membership: |
![]() ![]() |
