Assay Details
Gene Symbol: NSMCE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008270611.2 | XP_008268833.1 | 6 - 7 | 914 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NSMCE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008270611.2 | XP_008268833.1 | 6 - 7 | 914 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |