Assay Details
Gene Symbol: BICRAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008262921.2 | XP_008261143.1 | 9 - 10 | 2485 | 108 | |
XM_017345133.1 | XP_017200622.1 | 8 - 9 | 2364 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
