Assay Details
Gene Symbol: HPS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002721171.3 | XP_002721217.1 | 1744 | 60 | ||
XM_017350662.1 | XP_017206151.1 | 2060 | 60 | |||
XM_017350663.1 | XP_017206152.1 | 1368 | 60 | |||
XM_017350664.1 | XP_017206153.1 | 1821 | 60 | |||
XM_017350665.1 | XP_017206154.1 | 1684 | 60 | |||
XM_017350666.1 | XP_017206155.1 | 1952 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
