Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264899.2 | XP_008263121.1 | 2 - 3 | 357 | 76 | |
XM_008264900.2 | XP_008263122.1 | 3 - 4 | 955 | 76 | ||
XM_008264901.2 | XP_008263123.1 | 3 - 4 | 959 | 76 | ||
XM_008264902.2 | XP_008263124.1 | 2 - 3 | 353 | 76 | ||
XM_008264904.2 | XP_008263126.1 | 2 - 3 | 356 | 76 | ||
XM_008264905.2 | XP_008263127.1 | 2 - 3 | 358 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
