Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264899.2 | XP_008263121.1 | 6 - 7 | 4220 | 67 | |
XM_008264900.2 | XP_008263122.1 | 7 - 8 | 4818 | 67 | ||
XM_008264901.2 | XP_008263123.1 | 7 - 8 | 4822 | 67 | ||
XM_008264902.2 | XP_008263124.1 | 6 - 7 | 4216 | 67 | ||
XM_008264904.2 | XP_008263126.1 | 5 - 6 | 4096 | 67 | ||
XM_008264907.2 | XP_008263129.1 | 3 - 4 | 450 | 67 | ||
XM_008264909.2 | XP_008263131.1 | 3 - 4 | 451 | 67 | ||
XM_017346097.1 | XP_017201586.1 | 4 - 5 | 1052 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
