Assay Details
Gene Symbol: CMKLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249345.2 | XP_008247567.1 | 329 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008249345.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
