Assay Details
Gene Symbol: ERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008274888.2 | XP_008273110.1 | 514 | 58 | ||
XM_017350791.1 | XP_017206280.1 | 559 | 58 | |||
XM_017350792.1 | XP_017206281.1 | 559 | 58 | |||
XM_017350793.1 | XP_017206282.1 | 559 | 58 | |||
XM_017350794.1 | XP_017206283.1 | 393 | 58 | |||
XM_017350795.1 | XP_017206284.1 | 559 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
