Assay Details
Gene Symbol: NMNAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266242.2 | XP_008264464.1 | 4 - 5 | 571 | 72 | |
XM_008266243.2 | XP_008264465.1 | 4 - 5 | 567 | 72 | ||
XM_008266244.2 | XP_008264466.1 | 4 - 5 | 607 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
