Assay Details
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266580.2 | XP_008264802.2 | 8 - 9 | 1020 | 72 | |
XM_017346938.1 | XP_017202427.1 | 9 - 10 | 1297 | 72 | ||
XM_017346940.1 | XP_017202429.1 | 8 - 9 | 913 | 72 | ||
XM_017346941.1 | XP_017202430.1 | 7 - 8 | 887 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
