Assay Details
Gene Symbol: ATP5PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722947.3 | XP_002722993.1 | 149 | 46 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002722947.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
