Assay Details
Gene Symbol: COMMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709851.3 | XP_002709897.1 | 2 - 3 | 480 | 94 | |
XM_017340742.1 | XP_017196231.1 | 2 - 3 | 335 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
