Assay Details
Gene Symbol: MACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713706.3 | XP_002713752.2 | 3 - 4 | 2362 | 86 | |
XM_008261503.2 | XP_008259725.1 | 3 - 4 | 2281 | 86 | ||
XM_008261504.2 | XP_008259726.1 | 4 - 5 | 2304 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
