Assay Details
Gene Symbol: SERPINB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713632.3 | XP_002713678.1 | 6 - 7 | 872 | 74 | |
XM_017344224.1 | XP_017199713.1 | 6 - 7 | 868 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
