Assay Details
Gene Symbol: TOGARAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718163.3 | XP_002718209.1 | 3 - 4 | 2573 | 110 | |
XM_008269541.2 | XP_008267763.1 | 3 - 4 | 2574 | 110 | ||
XM_008269542.2 | XP_008267764.1 | 4 - 5 | 2521 | 110 | ||
XM_008269543.2 | XP_008267765.1 | 3 - 4 | 2574 | 110 | ||
XM_008269544.2 | XP_008267766.1 | 3 - 4 | 2574 | 110 | ||
XM_008269546.2 | XP_008267768.1 | 3 - 4 | 1658 | 110 | ||
XM_008269547.2 | XP_008267769.1 | 3 - 4 | 2575 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
