Assay Details
Gene Symbol: ZNF567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249567.2 | XP_008247789.1 | 889 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008249567.2 transcripts.
Set Membership: |
![]() ![]() |
