Assay Details
Gene Symbol: METTL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008250889.2 | XP_008249111.1 | 240 | 54 | ||
XM_017338911.1 | XP_017194400.1 | 240 | 54 |

More Information
Set Membership: |
![]() ![]() |
