Assay Details
Gene Symbol: NECTIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715216.3 | XP_002715262.1 | 2 - 3 | 729 | 118 | |
XM_002715217.3 | XP_002715263.1 | 2 - 3 | 728 | 118 | ||
XM_008264201.2 | XP_008262423.1 | 2 - 3 | 731 | 118 | ||
XM_008264202.2 | XP_008262424.1 | 2 - 3 | 729 | 118 | ||
XM_017345738.1 | XP_017201227.1 | 2 - 3 | 736 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
