Assay Details
Gene Symbol: NFATC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008252341.1 | XP_008250563.1 | 2112 | 54 | ||
XM_017339789.1 | XP_017195278.1 | 2112 | 54 | |||
XM_017339790.1 | XP_017195279.1 | 2112 | 54 | |||
XM_017339791.1 | XP_017195280.1 | 2112 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
