Assay Details
Gene Symbol: TAFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264651.2 | XP_008262873.1 | 1 - 2 | 369 | 70 | |
XM_017345967.1 | XP_017201456.1 | 1 - 2 | 379 | 70 | ||
XM_017345969.1 | XP_017201458.1 | 1 - 2 | 368 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008264651.2, XM_017345967.1, XM_017345969.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
