Assay Details
Gene Symbol: TAFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264651.2 | XP_008262873.1 | 4 - 5 | 730 | 90 | |
XM_017345967.1 | XP_017201456.1 | 4 - 5 | 740 | 90 | ||
XM_017345968.1 | XP_017201457.1 | 3 - 4 | 686 | 90 | ||
XM_017345969.1 | XP_017201458.1 | 4 - 5 | 690 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
