Assay Details
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267170.2 | XP_008265392.1 | 1 - 2 | 165 | 76 | |
XM_008267171.2 | XP_008265393.1 | 1 - 2 | 165 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008267170.2, XM_008267171.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
