Assay Details
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267170.2 | XP_008265392.1 | 2 - 3 | 246 | 78 | |
XM_008267172.2 | XP_008265394.1 | 2 - 3 | 187 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008267170.2, XM_008267172.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
